Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Kimtech™ M3 Sterile Face Masks
BIOPHARMAGREENER_CHOICE

Catalog No. 189992405 Shop All Ansell Products
Change view
Click to view available options
Fastening Type:
Earloops
Ties
Includes:
7 in. Soft Ties
9 in. Soft Ties
Knitted Earloops
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fastening Type Includes
18-999-2405 Ties 7 in. Soft Ties
18-999-4779 Earloops Knitted Earloops
18-999-4788 Ties 9 in. Soft Ties
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-999-2405 Supplier Ansell Supplier No. 62467
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.

  • Gamma-sterilized to 10-6
  • White with soft ties or earloops
  • Outer layer of apertured polyethylene film to provide protection from contamination
  • Polyester binding and polyester filter media provide bacteria filtration efficiency of 91.9% and particle filtration efficiency of 93.1%
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Material SMS
Size Standard
Sterility Sterile
Style Pleated
Cleanroom Class Class 1 (ISO 3)
Disposable Single-use
Filter Efficiency 89.3% (PFE), 94.1% (BFE)
Includes 7 in. Soft Ties
Latex Free Yes
Model 62467
Pore Size 0.1 μm (PFE), 3 μm (BFE)
Series Kimtech™ M3 Sterile
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.