Learn More
Ansell Kimtech™ M3 Sterile Face Masks
BIOPHARMAGREENER_CHOICE
Catalog No. 189994779 Shop All Ansell Products
Change view
Click to view available options
Fastening Type:
Earloops
Ties
Includes:
7 in. Soft Ties
9 in. Soft Ties
Knitted Earloops
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fastening Type Includes
18-999-4779 Earloops Knitted Earloops
18-999-2405 Ties 7 in. Soft Ties
18-999-4788 Ties 9 in. Soft Ties
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-999-4779 Supplier Ansell Supplier No. 62470
Add to Cart
Add to Cart
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
  • Gamma-sterilized to 10-6
  • White with soft ties or earloops
  • Outer layer of apertured polyethylene film to provide protection from contamination
  • Polyester binding and polyester filter media provide bacteria filtration efficiency of 91.9% and particle filtration efficiency of 93.1%
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Earloops
Material SMS
Size Standard
Sterility Sterile
Style Pleated
Cleanroom Class Class 1 (ISO 3)
Disposable Single-use
Filter Efficiency 89.3% (PFE), 94.1% (BFE)
Includes Knitted Earloops
Latex Free Yes
Model 62470
Pore Size 0.1 μm (PFE), 3 μm (BFE)
Series Kimtech™ M3 Sterile
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.