Learn More
Diagnostic Biosystems HEAT SHOCK PROTEIN 70 Heat Shock Protein 70 (HSP 70) (W27)
Supplier:  Diagnostic Biosystems MOB26905
Encompass_Preferred
This antibody is specific to constitutive (HSP73) and inducible (HSP72) form of HSP70. HSP70 is found to be associated with steroid receptors, actin, p53, polyoma T antigen, nucleotides and other known proteins. HSP70 has been shown to be involved in protective roles against thermal stress, cytotoxic drugs and other damaging conditions.
Catalog No. 50-238-5506
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.