Learn More
Thermo Scientific™ Heat-Shrink Bands for Nalgene™ PETG Media Bottles
GREENER_CHOICE
Catalog No. 0181453
Change view
Click to view available options
Neck Finish:
20-415
24-415
38-430
53B
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
0181453 53B
0181450 20-415
0181451 24-415
0181452 38-430
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0181453 Supplier Thermo Scientific™ Supplier No. 3121600530
Add to Cart
Add to Cart
Ensure the integrity of bottle contents with these tamper-resistant seals. Heat-shrink bands are gamma stable and include a perforated tear strip feature for easy removal.
Ensure the integrity of bottle contents with Thermo Scientific™ Heat-Shrink Bands for Thermo Scientific™ Nalgene™ PETG Media Bottles. Simply apply the recommended application torque for Thermo Scientific Nalgene closures and slide the heat-shrink band over the closure and bottle neck. Heat the band to shrink and secure contents with a tamper-resistant seal.
  • Gamma stable bands include a perforated ”tear strip” feature for easy removal.
  • Bands available are in four sizes to mate with Nalgene PETG Square Media Bottles.
  • Bands are packed 1,000 per case (two zipper bags of 500 each).
TRUSTED_SUSTAINABILITY
Specifications
Material PETG
No. per Case 1000
No. per Pack 1000
Torque 38-53 in.-lb
Torque Wrench Fittings 2195-1153
Neck Finish 53B
Product Type Heat-shrink Band
For Use With (Equipment) 2000 mL Square Media Bottle