Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Heat Transfer Apparatus
SDPSCI_ED

Catalog No. S00331 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S00331 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00331 Supplier United Scientific™ Supplier No. UNHTRF01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Heat Transfer Apparatus allows students to investigate the transfer of heat by conduction between two heat reservoirs.

  • The kit includes two plastic foam calorimeters with lids, a U-shaped aluminum bar which acts as a 'heat bridge', two digital thermometers for tracking the temperature changes in the masses of water held inside the calorimeters, and an activity guide
  • The calorimeters hold up to 200 mL of liquid and the digital thermometers measure temperatures between 50°C and 150°C with a precision of 0.1°C
  • The aluminum bridge has a cross sectional area of 122 mm2 and is about 200 mm long between the liquid reservoirs (the exact length can vary, depending on the volumes of water used)

Specifications

Grade Pre-K to 12
Format Kit
Product Type Heat Transfer Kit
Languages English
Subject Earth Science - Land, Water and Sea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.