Learn More
Eisco™ Heat Transfer Kit
SCI_ED
Catalog No. S12994 Shop All Eisco Products
Change view
Click to view available options
Grade:
7 to 12
Catalog No. Grade
S12994 7 to 12
1 options
Catalog No. S12994 Supplier Eisco™ Supplier No. PH1120144PK15
Add to Cart
Add to Cart
Demonstrate and learn about thermal energy and heat transfer. This Heat Transfer Kit introduce students to the concepts of thermal energy and heat transfer in a visually and kinesthetically effective manner.
  • Each of 15 kits contains: 2 Styrofoam calorimeters with lids, 1 aluminum heat transfer bar, 1 low range (-40°C to +50°C) thermometer (Plastic) and 1 high range (-30°C to +110°C) thermometer (metal)
  • Calorimeter dimensions: H x dia.: 3.75 x 3.5 in.
Specifications
Grade 7 to 12
Includes 30 Styrofoam Calorimeters with Lids, 15 Aluminum Heat Transfer Bars, 15 low Range Thermometers (Plastic), 15 High Range Thermometers (Metal)
Quantity 15 Pk.
Product Type Heat Transfer Kit