Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hughes Heated Mobile Self-Contained Emergency Safety Shower with Eye/Face Wash

Catalog No. 18666708 Shop All Justrite Safety Group Products
Change view
Click to view available options
Flow Rate:
3 gpm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
18-666-708 3 gpm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-666-708 Supplier Justrite Manufacturing Co Supplier No. H40K45GA1H

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Hughes Heated Mobile Self-Contained Emergency Safety Shower with Eye/Face Wash holds 30-gallons (114-liter) of water for use in responding to potential accidents anywhere. Suitable for temporary use.

Features:

  • Corrosion-resistant, polypropylene-lined pressurized cylinder and only requires the connection of a hose from a water main to fill it ready for use.
  • Ideal for cold climates with a minimum ambient operating temperature of -4°F (-20°C) or above
  • Trace tape heating with polyethylene jacket and polyurethane foam insulation improve frost protection and strength.
  • Easily maneuvered by one person
  • Incorporates a stainless steel frame and large diameter pneumatic tires—adjustable wheel spacing allows the unit to go through standard door openings.
  • Built-in A-frame with towing hitch makes it simple to move longer distances or over uneven surfaces by using any small vehicle fitted with a standard tow ball.
  • Disconnect shower arm for compact transportation.
  • High flow rate features an integral lid connected to the water valve for automatic operation—activate by simply pulling down the lid.
  • Easy-to-reach flow regulator valve positioned on the side of the bowl adjusts to suit the severity of the accident.
  • Suitable for Class 1 Division 2 hazardous areas, as well as non-hazardous areas.
  • For cold climates with a minimum ambient operating temperature of -4°F (-20°C) or above
  • Mobile eye/face wash and shower provides a continuous flow of water for 1-1/2 minutes
  • High-performance quarter-turn ball valves made of 316L stainless steel endure the harshest environments and site conditions
  • Covered, durable acrylic-capped ABS eye/face wash protects the bowl and eyewash diffusers from dust and debris
  • Twin diffusers provide a gentle flow of aerated water and prevent further injury to the eyes

Compliance

Not ANSI Compliant

Warranty and Services

1 Year

Specifications

Activation Hand Lever: Pull Lid
Bowl Size (Metric) 241 mm
Capacity (English) 30 gal.
Color White with Green Labels
Depth (English) 72 in.
Depth (Metric) 1829 mm
Electrical Requirements 120 V
Flow Rate 3 gpm
For Use With (Application) Emergency Shower and Eye Wash with Water
Height (English) 60 in.
Height (Metric) 152.4 cm
Includes 1 Unit
Inlet Connection (Metric) 0.5 in. NPT Coupling
Material Polypropylene, Stainless Steel, Chrome Plated Brass, Nylon 6 Red
Material (Piping Connection) Stainless Steel
Mounting Portable
Pressure 29 to 87 psi
Product Type Safety Shower and Emergency Eye Wash
Width (English) 45 in.
Width (Metric) 1143 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.