Learn More
Grainger BRISKHEAT Heating Cord, 12FT 120V 144WATT
Supplier:  Grainger 36TW86
Encompass_Preferred
The BriskHeat Heating Cord is a 12 foot long heating cord that provides 144 watts of power. It has a maximum continuous exposure temperature of 65°C (149°F) and is made from fiberglass yarn, silicone rubber and tinned copper bus wires. This heating cord can be used for pipe tracing, gutter de-icing, under sink heating, wrapping valves and more. It comes with a 3 prong plug for easy power connection. The cord has been agency approved for safety.
Catalog No. NC2862932