Learn More
Chemglass Life Sciences 100mL Glas-Col Heating Mantle, Series inSTMin, Spherical, 230 Watts
Supplier: Chemglass Life Sciences CG1250001
100mL Glas-Col Heating Mantle, Series STM, Spherical, 230 Watts, The Glas-Col Series STM mantle performs the same function as a Series TM heating mantle, plus operations requiring more heat. This mantle is useful in distillation and fractionation processes, especially when time is important. The higher temperature range enables operations to be performed that require more heat, such as rapid distillation of water and liquid-phase cracking in a flask, where the products are taken out of the flask through a column. Withstanding an upper internal operating temperature of 650 C, the STM mantle is very resistant to burnout. The STM mantle has a durable aluminum exterior and a resilient and pliable interior to cradle glassware. A splash guard and poncho safety shield can be used to prevent damage from accidental spills.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.