Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100A O398
Encompass_Preferred
Hemispherical heating mantle for 125ml single or multi neck flasks to 400 degrees C maximum internal operating temperature, 80W, 115V, CSA.
Catalog No. NC0821676
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.