Learn More
Description
- Vented for better gas exchange
- Heavy design for heat resistance using hot agar
- Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications
Specifications
| Color | Clear |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Non-sterile |
| Type | Petri Dish |
| No. per Case | 480 |
| Vent | Yes |
| Height (Metric) | 16 mm |
| Diameter (Metric) | 94 mm |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.