Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes

Catalog No. 07000323 Shop All Greiner Bio-One Products
Change view
Click to view available options
Diameter (Metric):
94 mm
100 mm
Height (Metric):
16 mm
20 mm
Sterility:
Non-sterile
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Height (Metric) Sterility
07-000-323 94 mm 16 mm Sterile
07-000-326 100 mm 20 mm Non-sterile
07-000-321 94 mm 16 mm Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-323 Supplier Greiner Bio-One Supplier No. 633161

Used for easy stacking. Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes is designed for better heat resistance using hot agar.

  • Vented for better gas exchange
  • Heavy design for heat resistance using hot agar
  • Manufactured from crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Polystyrene
Shape Round
Sterility Sterile
Type Petri Dish
No. per Case 480
Vent Yes
Height (Metric) 16 mm
Diameter (Metric) 94 mm

For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.