Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Heavy-Duty Polypropylene Coveralls, CVL-NW-HD-E
SDP

Catalog No. 19055276
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Color:
Blue
White
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-055-276 4X-Large White
19-055-278 Medium White
19-055-269 Medium Blue
19-055-279 Large White
19-055-270 Large Blue
19-055-280 X-Large White
19-055-271 X-Large Blue
19-055-281 2X-Large White
19-055-272 2X-Large Blue
19-055-282 3X-Large White
19-055-273 3X-Large Blue
19-055-274 4X-Large Blue
19-055-277 5X-Large White
19-055-275 5X-Large Blue
19-070-136 6X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 19-055-276 Supplier Keystone Safety Supplier No. CVLNWHDE4XLWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Heavyweight polypropylene coveralls are made of breathable fabric that provides economical protection against dry particulates, dirt, and grime. Available in blue or white.

  • Heavyweight polypropylene coveralls
  • Breathable fabric provides economical protection
  • Elastic wrists and ankles
  • Low cost solution to protect against dry particulates, dirt, and grime
  • 100% latex free

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material Polypropylene (Heavy-duty)
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Certifications/Compliance Cleanroom Class 7
Model CVL-NW-HD-E-LRG
No. of Pockets None
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.