Learn More
Ansell HyFlex™ 70-225 Medium Duty String Knit Gloves with Aramid Fiber
Catalog No. 19042205 Shop All Ansell Products
Change view
Click to view available options
Size:
9
Catalog No. Size
19-042205 9
1 options
This item is not returnable. View return policy
Catalog No. 19-042205 Supplier Ansell Supplier No. 222127
  • $6,710.79
    $3,053.65 / Case of 144
    Save $3,657.14
    54% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Aramid fiber medium duty industrial gloves offers high cut-resistance to weight ratio. Ideal for automotive or machinery and equipment industries.
  • Five times the cut resistance of leather
  • Heat protection
  • Hand-to-hand reversible
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Neptune
Color Yellow
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Latex Free Yes
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size 9
Glove Cuff Style Knit
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.