Learn More
Enzo Life Sciences Hecameg (1g). CAS: 115457-83-5
Supplier: Enzo Life Sciences ALX500003G001
Non-ionic, glucose based very mild detergent for membrane protein purification. Alternative name: 6-O-(N-Heptylcarbamoyl)-methyl-alpha-D-glucopyranoside. Purity: ≥98% (HPLC). Solubility: Soluble in water (>100mg/ml at +4°C). Long Term Storage: +4°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.