Learn More
AdipoGen Hederacoside C (50 mg)
SDP
Supplier:  AdipoGen CDXH0299M050
Encompass_Preferred
Chemical. CAS: 14216-03-6. Formula: C59H96O26. MW: 1221.38. Hederacoside C is a major bioactive constituent isolated from Hedera helix L. It exhibits biological properties such as antibacterial, expectorant, bronchodilator, bronchospasmolytic and anti-inflammatroy effects. It is widely used in the treatment of respiratory disorders involving acute respiratory infections, chronic inflammatory bronchitis and productive coughs. It reduces TNF-alpha, IL-1beta, IL-6, COX-2, and nitric oxide synthase (NOS) levels in a concentration-dependent manner and IL-1 receptor-associated kinase-1 (IRAK1) activity. It reduces scopolamine-induced memory impairment. Hederacoside C markedly suppressed TLR2 and TLR4 expressions by attenuating the MAPKs (p38, ERK, JNK) and NF-kappa (p65 and IkappaBalpha) pathways followed by decreasing the phosphorylation of p38, ERK, JNK, p65, and IkappaBalpha.
Catalog No. 50-205-4501
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.