Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hegsted Salt Mixture

Hegsted Salt Mixture is used for rodent diet.

$180.00 - $3273.00

Specifications

Product Type Animal Diet
For Use With (Application) Rodent diet
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP290284002
View Documents
MP Biomedicals™
0290284002
2 kg
Each for $180.00
Only null left
Add to Cart
 
MP290284010
View Documents
MP Biomedicals™
0290284010
10 kg
Each for $679.00
Only null left
Add to Cart
 
MP290284050
View Documents
MP Biomedicals™
0290284050
50 kg
Each for $3,273.00
Only null left
Add to Cart
 
Description

Description

  • Mineral Mix
  • For rodent diet
  • Powder
Specifications

Specifications

Animal Diet
Rodent diet
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.