Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Evaporating Flasks
SDP
Evaporating flasks for use with Heidolph Hei-VAP series rotary evaporators
Supplier:  Heidolph™ 036302320
 View more versions of this product
Catalog No. 13-878-689
Only null left
Add to Cart
Description
Description
  • Joint Size options of: NS 29/32 or NS 24/40
Specifications
Specifications
Evaporating Flask
Hei-VAP Rotary Evaporator
29/32
250 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.