Learn More
Heidolph™ Glassware - Dry Ice Condenser Jacket
SDP
Catalog No. 13878624 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Application):
For Low-Boiling Solvents
Catalog No. For Use With (Application)
13-878-624 For Low-Boiling Solvents
1 options
Catalog No. 13-878-624 Supplier Heidolph™ Supplier No. 036301670
Add to Cart
Add to Cart
For low-boiling solvents
  • Halar-coated for added safety
Specifications
For Use With (Application) For Low-Boiling Solvents
Type Condensate Trap
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov