Learn More
Heidolph™ Glassware - G3 Glass Set
SDP
Catalog No. 13878598 Shop All Heidolph North America Products
Change view
Click to view available options
Standard Taper Joint No:
24/40
Catalog No. Standard Taper Joint No
13-878-598 24/40
1 options
Catalog No. 13-878-598 Supplier Heidolph™ Supplier No. 036301410
Add to Cart
Add to Cart
Vertical condenser for all standard distillations
Vertical condenser for all standard distillations; the option with the smallest space requirement.
  • Glassware set includes on 1,000ml evaporating flask and one 1,000ml receiving flask
  • Evaporating flasks and vapor tubes which come with a standard NS 24/40 joint size are also available with NS 24/32 ground joints
  • Allows for the automated distillation program AUTO accurate on Hei-VAP Precision models
  • All glassware sets feature GL 10 thread; the inlet tube can be connected with a threaded fitting
Specifications
For Use With (Application) For all standard distillations, the option with the smallest space requirement
Standard Taper Joint No 24/40
Type Vertical Condenser
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov