Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Vacuum Valve
SDP
Required to control vacuum for Hei-VAP models with valve-regulated pumps
Supplier:  Heidolph™ 036308070
Catalog No. 13-878-534
This item is not returnable. View return policy
Description
Description
  • External attachment design for easy cleaning
  • Can be combined with the Woulff Bottle to one unit
  • Not required if you utilize a manual vacuum controller
Specifications
Specifications
Vacuum Valve
Hei-VAP rotary evaporator
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.