Learn More
Heidolph™ Hei-Vap Rotary Evaporator Vapor Tubes
SDP
Accessory parts and components for Heidolph Hei-VAP Rotary Evaporators
$516.50
Specifications
Type Vapor Tube
For Use With (Equipment) Hei-VAP Rotary Evaporator
Includes Vapor tube only. (Clamping sleeve is sold separately; Heidolph No. 036308030.)
View More Specs
Includes: Vapor tube only. (Clamping sleeve is sold separately.)
Products 2
Catalog Number Mfr. No. Taper Price Quantity & Availability  
Catalog Number Mfr. No. Taper Price Quantity & Availability  
13-878-568
SDP
View Documents
Heidolph™
036301110
24/40
Each for $516.50
Only null left
Add to Cart
13-878-567
SDP
View Documents
Heidolph™
036301100
29/32
Each for $516.50
Only null left
Add to Cart
Description
Description
  • The patented clamping sleeve system allows the removal of the vapor tube easily from the drive
  • Years of performance life reduce your spare parts budget up to 75%
  • The Hei-VAP vapor tube does not get stuck in the drive, thus eliminating the risk of broken glass
  • Immediate removal of the vapor tube from the drive facilitates your work
Specifications
Specifications
Vapor Tube
Hei-VAP Rotary Evaporator
Vapor tube only. (Clamping sleeve is sold separately; Heidolph No. 036308030.)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.