Learn More
Heidolph™ Overhead Stirrer Accessory, Stir Shafts
SDP
Catalog No. 05405294 Shop All Heidolph North America Products
Change view
Click to view available options
Shape:
Anchor
Retreat Curve
Turbine
Model:
015950137
015950138
015950139
015950140
015950141
015950142
015950143
015950144
015950145
015950146
015950148
015950150
015950151
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shape Model
05-405-294 Retreat Curve 015950150
05-405-281 Anchor 015950137
05-405-282 Retreat Curve 015950138
05-405-283 Turbine 015950139
05-405-286 Turbine 015950142
05-405-284 Anchor 015950140
05-405-288 Retreat Curve 015950144
05-405-289 Turbine 015950145
05-405-287 Anchor 015950143
05-405-285 Retreat Curve 015950141
05-405-290 Anchor 015950146
05-405-291 Retreat Curve 015950146
05-405-292 Turbine 015950148
05-405-295 Turbine 015950151
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 05-405-294 Supplier Heidolph™ Supplier No. 015950150
Add to Cart
Add to Cart
Stir shafts for use with various Heidolph overhead or magnetic stirrers
For use with overhead stirrers
PTFE
Various blade styles to choose from
Specifications
For Use With (Equipment) Overhead Stirrer
Model 015950150
Shape Retreat Curve
Type Stirrer Shaft