Description
- Clamps provide different configurations depending on Perforated Platform and flask size used
Specifications
Specifications
| Holds | 1000 mL Erlenmeyer Flask |
| Product Type | Flask Clamp |
| For Use With (Equipment) | 1000 mL Erlenmeyer Flask of Perforated Platform 100, 1000 and 2000 Platform Shakers |
| Size | 6 |