Description
- Features Rodaviss joint and is compatible with Reactor-Ready lids
Specifications
Specifications
| Product Type | Reducing Adapter |
| Standard Taper Joint No. | B29 to B19 |
| Thread Size | B29 to B19 |