Learn More
Heidolph™ Radleys Enlarging Adapter and Reducing Adapter
SDP
Catalog No. 05405316 Shop All Heidolph North America Products
Change view
Click to view available options
Thread Size:
B24 to B19
B24 to B29
B29 to B19
B29 to B24
B29 to B34
B34 to B19
B34 to B24
B34 to B29
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size
05-405-316 B34 to B19
05-405-311 B24 to B29
05-405-312 B29 to B34
05-405-313 B24 to B19
05-405-315 B29 to B24
05-405-314 B29 to B19
05-405-317 B34 to B24
05-405-318 B34 to B29
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 05-405-316 Supplier Heidolph™ Supplier No. 015950174
Add to Cart
Add to Cart
Enlarging adapter and reducing adapter for use with Reactor-Ready™ lab reactors
  • Features Rodaviss joint and is compatible with Reactor-Ready lids
Specifications
Product Type Reducing Adapter
Standard Taper Joint No. B34 to B19
Thread Size B34 to B19
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov