Learn More
Heidolph™ Radleys GreenHouse Blowdown Evaporator™ Accessories - Vial Racks
Shop All Heidolph North America Products
Description
A selection of vial racks are available to hold the most popular vial sizes in 6 x 4 and 4 x 2 arrays
Specifications
Specifications
| For Use With (Equipment) | Heidolph Radleys GreenHouse Blowdown Evaporator |
| Holds | 6 x 4, for 13.8 mm OD Vials |
| Type | AT03 Rack |