Learn More
Heidolph™ Radleys™ Reaction Vessel Accessory: DN200 Lids
Glass lids with Rodaviss conical ground glass joints
$3285.70 - $3830.57
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
13-890-015
![]() |
Heidolph™
015908005 |
6-Neck Lid |
Each for $3,285.70
|
|
|||||
13-890-016
![]() |
Heidolph™
015908006 |
7-Neck Lid |
Each for $3,830.57
|
|
|||||
Description
- For reaction vessels used in Reactor-Ready™ Pilot Lab Reactor
- Lid with grease free Rodaviss joints that are interchangeable with ordinary 1-in-10 B and A length tapered conical joints and will not jam
- Available as 6 or 7 neck
Inlets in 6 Neck DN200 Lid
- 1 x B34 central vertical - stirrer
- 1 x B19 side vertical - probe
- 1 x B24 side vertical - gas purge
- 1 x B29 side vertical - dropping funnel, bubbler
- 1 x B34 side vertical - condenser and reflux
- 1 x B45 side angled - solid additions
Inlets in 7 Neck DN200 Lid
- 1 x B34 central vertical - stirrer
- 1 x B19 side vertical - probe
- 1 x B24 side vertical - gas purge
- 1 x B29 side vertical - drop funnel, bubbler
- 2 x B34 side vertical - condenser and reflux
- 1 x B45 side angled - solid additions
Specifications
Reaction Vessel |