Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Radleys SpeediFlow™ Adapter Heads
SDP

Catalog No. p-4835750 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
70 mL Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-403-733 70 mL Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-403-733 Supplier Heidolph™ Supplier No. 015860040
Only null left
Add to Cart
Add to Cart
Request a Quote

SpeediFlow™ adapter heads for use with Carousel Work-up Station™

  • The SpeediFlow is a hand held pressurizer which fits into the top of your plastic column, to push the mobile phase through the column.
  • Features a mechanically expandable Viton O-ring, operated by a trigger with an integral locking device, for unsupported gas tight operation
  • SpeediFlow features a safety relief valve that vents at 6 to 8 psi, preventing accidental over-pressure
  • SpeediFlow Booster increases flow rates to speed up your work-ups.
  • The SpeediFlow includes a range of optional adapters to fit all standard plastic column sizes including 15 ml, 25 ml, 70 ml, and 150 ml, plus the Whatman AutoCup™. Thus allowing the SpeediFlow to be used in other SPE or filtration applications.
  • Specifications

    Type SpeediFlow
    For Use With (Equipment) 70 mL Columns
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.