Learn More
Heidolph™ Stirrer Chucks for RZR Overhead Stirrer
SDP
Catalog No. 13880151 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
2102 Z Stirrer
RZR 1 Stirrer
RZR 2020 - 2102 Stirrers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-880-151 RZR 2020 - 2102 Stirrers
13-880-150 RZR 1 Stirrer
13-889-463 2102 Z Stirrer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-880-151 Supplier Heidolph™ Supplier No. 036300570
Accessories for use with RZR Overhead Stirrers
13880150
  • For use with RZR 1 stirrer
  • 8 mm
13880151
  • For use with RZR 2020 - 2102 stirrers
  • 10.5 mm
13889463
  • For use with 2102 Z stirrer
  • 10.5 mm
Specifications
Diameter (English) Shaft 0.39 in.
Diameter (Metric) Shaft 1 cm
For Use With (Equipment) RZR 2020 - 2102 Stirrers
Type Stirrer Chuck
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov