Learn More
Seca Corp 220 Telescopic measuring rod for seca column scales, inches/cm
Supplier: Seca Corp 2201814003
This telescopic measuring rod can be fitted at any time to many seca column scales. It has the same convenient eye-level read out for even taller patients. • Telescopic measuring rod accessory for seca column scale models 700, 703, 769 • Convenient eye-level read out for even taller patients • Measuring range: 24 – 78 in / 60 – 200 cm • Graduation: 1/8 in / 1 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.