Learn More
Description
This antibody is designed to detect the protein of the HEK293 cells.
Specifications
Specifications
| Antigen | HEK 293 cells |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene Alias | 293, HEK 293 cells, HEK293 |
| Host Species | Rabbit |
| Immunogen | Peptide |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
