Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEK 293T cells Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7212609 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5698 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5698 Supplier Proteintech Group Inc Supplier No. 275551AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is designed to detect the proteins of the HEK 293T cells.

HEK 293T cell line, an important variant of HEK 293 cell line, which contains the SV40 Large T-antigen that allows for episomal replication of transfected plasmids containing the SV40 origin of replication. HEK 293T cells are usually used for plasmids transfection and temporal expression of desired gene products.

Specifications

Antigen HEK 293T cells
Applications ELISA
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene Alias 293T, HEK 293T cells, HEK293T
Host Species Rabbit
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.