Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam HeLa Membrane Extract Lysate, Liquid, WB, 100 μg

Catalog No. 502678649
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-267-8649 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-267-8649 Supplier Abcam Supplier No. AB29547100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HeLa membrane extract lysate suitable for Western blot analysis.

  • Applications: WB
  • Form: Liquid
  • Storage buffer constituents: HEPES, Magnesium chloride, Potassium chloride, Sucrose, 10% Glycerol, 2% Sodium lauryl sulfate, 1.2% 2-Mercaptoethanol, 1% 4-Nonylphenol, branched, ethoxylated, and 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Human
Form Liquid
Detection Location In-Blot Detection
Buffer HEPES, magnesium chloride, potassium chloride, sucrose, 10% glycerol, 2% sodium lauryl sulfate, 1.2% 2-mercaptoethanol, 1% 4-nonylphenol, branched, ethoxylated, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione
Cross Reactivity Human
Formulation HEPES, Magnesium chloride, Potassium chloride, Sucrose, 10% Glycerol, 2% Sodium lauryl sulfate, 1.2% 2-Mercaptoethanol, 1% 4-Nonylphenol, branched, ethoxylated, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione
Sample Type HeLa cells
Quantity 100 μg
For Use With (Application) Western Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.