Learn More
Active Motif HeLa nuclear extract
SDP
Supplier:  Active Motif 36010
Encompass
Active Motif's HeLa nuclear extract was prepared from a cell culture of the HeLa human epithelial carcinoma cell line. The HeLa cell line was originally derived from cells obtained in 1951 from Henrietta Lacks, a cervical cancer patient who eventually died from her cancer. The HeLa cell line is one of the oldest and most commonly utilized immortal cell lines in scientific research. These cells are highly prolific and easily sustainable, allowing for flexibility in their application in various fields of scientific research, including cancer biology, virology, and human disease. To date, over 60,000 publications across various disciplines have referenced the use of HeLa cells.
Catalog No. NC1361090
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.