Learn More
Diagnostic Biosystems HELICOBACTER PYLORI Helicobacter Pylori Antibody
Supplier:  Diagnostic Biosystems RP16901
Encompass_Preferred
This antibody reacts with the 54 kDa (flagellin) of H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.
Catalog No. 50-238-5350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.