Learn More
ATCC Helicobacter pylori (Marshall et al.) Goodwin et al. - ATCC 43526 Frozen culture
Supplier: ATCC 43526
Helicobacter pylori (Marshall et al.) Goodwin et al. (ATCC 43526) is a clinical research strain supplied as a frozen culture for laboratory investigations of H. pylori physiology, virulence determinants, and antimicrobial testing. It requires specialized microaerophilic conditions and handling under appropriate biosafety practices.
- Slow-growing; requires moist growth conditions.
- Recommended media: trypticase soy agar or broth; blood-supplemented agar for enhanced growth.
- Incubation at 37°C under microaerophilic atmosphere (3-5% O2, 10% CO2).
- Provided as a frozen culture; store at -80°C or colder.
- For long-term preservation, freeze with 20% sterile glycerol prepared from pooled broth.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.