Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Helicobacter Pylori OMP Recombinant Rabbit Monoclonal Antibody (RM411)
GREENER_CHOICE

Catalog No. PIMA544573
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA544573 Supplier Invitrogen™ Supplier No. MA544573
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody reacts to Histone H2A.X acetylated at Lysine 9 (K9ac). No cross reactivity with non-modified Lysine 9 or other acetylated Lysines in histone H2A.

Outer membrane proteins (OMPs) ofH. pyloriplay a crucial role in the host pathogen interaction, virulence and pathogenesis. When OMPs interact with the host immune cells, OMPs stimulate the production of pro-inflammatory molecules that may result in an uncontrolled inflammation leading to the transformation of a normal cell to a cancerous cell. The Helicobacter outer membrane family of OMPs inH. pyloriconsists of four members (HomA, B, C and D).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Helicobacter Pylori OMP
Applications ELISA
Classification Recombinant Monoclonal
Clone RM411
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Host Species Rabbit
Immunogen Proteins purified from Helicobacter pylori which correspond to the full length protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.