Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helios Hamster Anti-Human, Oligo, Clone: 22F6, BD AbSeq™

Catalog No. BDB667398 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB667398 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB667398 Supplier BD Biosciences Supplier No. 940509
Add to Cart
Add to Cart

Hamster Oligoclonal Antibody

Helios Oligoclonal antibody specifically detects Helios in Human,Mouse (Tested in Development) samples. It is validated for Single Cell 3'Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen Helios
Applications Single Cell 3' Sequencing
Classification Oligoclonal
Clone 22F6
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias IKAROS family zinc finger protein 2; ANF1A2; HELIOS; ZNF1A2; ZNFN1A2
Host Species Hamster
Immunogen Mouse Helios Peptide
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22779, 22807
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.