Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HELIOS Monoclonal Antibody (22F6), PE-Cyanine5, eBioscience™

Catalog No. 15988342
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-988-342 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-988-342 Supplier Invitrogen™ Supplier No. 15988342
Only null left
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: This 22F6 monoclonal antibody reacts with human, mouse and canine Helios. Helios is a zinc-finger transcription factor first identified for its similarity to Ikaros. It is expressed in hematopoietic stem cells in both the embryo and adult mice. While the highest expression is found in regulatory T cells, Helios is not expressed in mature B cells, dendritic cells or myeloid cells. Helios can form a homodimer as well as heterodimerize with other Ikaros family members to regulate transcription. Eight isoforms of Helios exist, some of which possess a dominant-negative function that has also been found to be upregulated in lymphoid malignancies, including T-acute lymphoblastic leukemia. 22F6 has been reported to cross react with rat Helios. Applications Reported: This 22F6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 22F6 antibody has been pre-diluted and tested by flow cytometric analysis of mouse lymph node cells using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please refer to 'Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins' located at www.thermofisher.com/flowprotocols . This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to...

This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HELIOS
Applications Flow Cytometry
Classification Monoclonal
Clone 22F6
Concentration 5 μL/Test
Conjugate PE-Cyanine5
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IKZF2
Gene Accession No. P81183, Q9UKS7
Gene Alias A730095J18Rik; ANF1A2; Helios; IKAROS family zinc finger 2; IKAROS family zinc finger 2 (Helios); ikaros family zinc finger protein 2; Ikzf2; Zfpn1a2; zinc finger DNA binding protein Helios; zinc finger protein Helios; zinc finger protein, subfamily 1A, 2 (Helios); ZNF1A2; Znfn1a2
Gene Symbols IKZF2
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22779, 22807, 488510
Target Species Canine, Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.