Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Helium-Hydrogen Spectrum Tube
SDPSCI_ED

Catalog No. S27846 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
8 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27846 8 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27846 Supplier United Scientific Supplies Supplier No. SPT2HH1
Add to Cart
Add to Cart

Standard Spectrum Tube filled with a mixture of hydrogen and helium

  • Explore star composition and observe behaviors of hydrogen and helium
  • An electrode is sealed in each end and attached to a looped metal cap for convenient hookup to wire connections
  • Activity guide included

Specifications

Color Silver
Grade 8 to 12
Age 13 to 18
For Use With (Application) Exploring Star Composition
Length (Metric) 25 cm
Length (Metric) Capillary 10 cm
Product Type Spectrum Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.