Learn More
MCR Safety HellKat 2 MAX6 Lens
Catalog No. 19812657 Shop All MCR Safety Products
Change view
Click to view available options
Lens Tint:
Amber
Blue
Brown
Clear
Gray
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-812-657 Blue
19-812-658 Gray
19-812-659 Clear
19-812-655 Brown
19-812-656 Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-657 Supplier MCR Safety Supplier No. HK213PF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
HellKat 2 MAX6 Lens gives you all the features you want for a comfortable and stylish option in protective eyewear. This generous lens offers an outstanding orbital seal and prevents hazards from threatening one of your most treasured belongings - your eyesight.
  • Lightweight, contoured frameless design
  • MAX6™ premium anti-fog lens coating
  • Excellent orbital seal for eye protection
  • Non-slip soft bayonet temple
  • Soft, secure TPR nose pad
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Lens Tint Blue
For Use With (Application) Manufacturing
Frame Color Blue
No. per Pack 1 Pair
Lens Material Polycarbonate
Frame Material Polycarbonate