Description
- Lightweight, contoured frameless design
- MAX6™ premium anti-fog lens coating
- Excellent orbital seal for eye protection
- Non-slip soft bayonet temple
- Soft, secure TPR nose pad
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Specifications
| Lens Tint | Gray |
| For Use With (Application) | Manufacturing |
| Frame Color | Gray |
| No. per Pack | 1 Pair |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |