Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HELLS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100261 Supplier Invitrogen™ Supplier No. PA5100261
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HELLS.

This gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HELLS
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HELLS
Gene Accession No. Q60848, Q9NRZ9
Gene Alias AI323785; E130115I21Rik; helicase, lymphoid specific; helicase, lymphoid-specific; hells; ICF4; LSH; Lymphocyte-specific helicase; lymphoid-specific helicase; Lysh; Nbla10143; PASG; Proliferation-associated SNF2-like protein; SMARCA6; SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6; SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6; Tbc1d12; TBC1D12: TBC1 domain family, member 12; YFK8
Gene Symbols HELLS
Host Species Rabbit
Immunogen A synthesized peptide derived from human HELLS(Accession Q9NRZ9), corresponding to amino acid residues Y480-K530.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15201, 294071, 3070
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.