Learn More
Brady™ Helvetica™ Quik-Align™ Lower Case Kit
Catalog No. 19108143 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on Blue
White on Blue
Height (English):
2 in.
3 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Height (English)
19-108-143 Black on Blue 3 in.
19-108-142 Black on Blue 2 in.
19-108-144 White on Blue 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-108-143 Supplier Brady™ Supplier No. 80013
Labels identify equipment, bins and areas of the warehouse required by facility safety and maintenance. Brady™ Helvetica Quik-Align™ Lower Case Kit are accurate reproductions of Helvetica letter style and has become the international standard for modern identification.
  • Made of pressure sensitive matte finish B-933 vinyl
  • Pigmented throughout for durability and fade resistance
  • Characters are located on notched carrier cards that interlock to provide “automatic” alignment and interletters spacing
Specifications
Includes Lower case a thru z legends
Material Vinyl
Adhesive Type Permanent Acrylic
Description Vinyl (B-933); Die-Cut, Quik-Align
Height (English) 3 in.
Product Type Lower Case Kit
Color Black on Blue