Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Hemastix, 50/Vial
SCI_ED

Catalog No. S80808 Shop All Eisco Products
Change view
Click to view available options
Tip Type:
Regular
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Type
S80808 Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80808 Supplier Eisco™ Supplier No. BI02017006
Add to Cart
Add to Cart

Vial of 50 dip-and-read Hemastix test strips for detecting the presence of blood in urine or at crime scenes.

  • Dip and read test for blood in urine.
  • Can be used at crime scenes to confirm suspected blood evidence.
  • Simply rub the Hemastix on a moistened stain or across a swab that has a suspected dried blood sample on it.
  • The reagent coated tip will turn a shade of green if hemoglobin is detected.
  • Vial of 50 strips.

Specifications

Handle Material Plastic
Tip Type Regular
Shelf Life 1 Year
Format Test Strips
Sterility Non-sterile
Certifications/Compliance None
Cap Color White
Product Type Collection Swabs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.