Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematein 94%, Thermo Scientific™

Catalog No. AC411150250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC411150250 25 g
AC411150050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411150250 Supplier Thermo Scientific Chemicals Supplier No. 411150250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 475-25-2
Molecular Formula C16H12O6
Molecular Weight (g/mol) 300.27
MDL Number MFCD00036187
InChI Key HLUCICHZHWJHLL-UHFFFAOYNA-N
Synonym hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein
PubChem CID 10138
IUPAC Name 5,6,10,15-tetrahydroxy-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-1(17),2,4,6,12,15-hexaen-14-one
SMILES OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O

Specifications

Color Brown
Infrared Spectrum Authentic
Quantity 25 g
Merck Index 15, 4669
Formula Weight 300.26
Physical Form Crystalline Powder
Chemical Name or Material Hematein, 94%
EINECSNumber 207-492-8
RTECSNumber DE3120000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.