Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Hematology Phosphate Buffer

Catalog No. 22110656 Shop All Epredia Products
Change view
Click to view available options
pH:
6.6
7.2
Quantity:
9.25 g
12 x 9.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Quantity
22-110-656 7.2 12 x 9.25g
22-050-294 6.6 9.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-110-656 Supplier Epredia™ Supplier No. 89033

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Combine Epredia™ Phosphate Buffer with Giemsa, Wright, and Wright-Giemsa stains for excellent results.

Supplied as 9.25g powder.

Specifications

Product Type Phosphate Buffered Saline (PBS)
Quantity 12 x 9.25g
pH 7.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.