Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Hematopoietic Colony Forming Cell Assay Kit, Fluorescent, 1x96 Tests

Catalog No. 502704230
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4230 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4230 Supplier Abcam Supplier No. AB2422881X96TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent assay kit for the detection of hematopoietic colony forming cells.

  • Detection method: Fluorescent

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Hematopoietic Colony Forming Cell
Format 96-well Plate Kit
No. of Reactions 96 Tests
Detection Method Fluorescence
Quantity 96 Tests
For Use With (Application) Hematopoietic Colony Forming Cell Assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.