Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Hematoxylin Counterstain
SDP

Catalog No. NB398961 Shop All Novus Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398961 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398961 Supplier R&D Systems™ Supplier No. H3401500NB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. based on Gill’s formulation, it produces blue stained nuclei with crisp detail.

  • Produces blue-stained nuclei with crisp detail and excellent color contrast.
  • Alcohol- and mercury-free formulation for safer laboratory use.
  • Compatible with most peroxidase and alkaline phosphatase substrates.
  • Suitable for use with both aqueous and non-aqueous mounting media.
  • Designed for immunohistochemistry (IHC), immunocytochemistry/immunofluorescence (ICC/IF), and in situ hybridization (ISH) applications.
  • Supplied in a ready-to-use format for convenient laboratory workflows.
  • Store at 15°C to 25°C (room temperature).

Specifications

For Use With (Application) Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, In-Situ Hybridization
Product Line Novus
Quantity 500 mL
Storage Requirements Room temperature
Product Type Counterstain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.